Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenol, 98+%

Catalog No. p-7082008
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1223622 100 g
AAA1223636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1223622 Supplier Thermo Scientific Chemicals Supplier No. A1223622
Only null left
Add to Cart

CAS: 108-68-9 | C8H10O | 122.17 g/mol

3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).

Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.

Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

Melting Point 61°C to 65°C
Density 1.12
Boiling Point 222°C
Flash Point 109°C (228°F)
Quantity 100 g
UN Number UN2261
Beilstein 774117
Merck Index 14,10082
Solubility Information Soluble in ethyl alcohol,carbon tetrachloride,benzene,chloroform,water and ether.
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylphenol
Show More Show Less
Hazard Category H301+H311-H314
Hazard Statement GHS H Statement
H301-H311-H314
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
EINECSNumber 203-606-5
RTECSNumber ZE6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.