Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. D34745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34745G 5 g
D347425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34745G Supplier TCI America Supplier No. D34745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 54132-75-1 | C9H9NO | 147.177 g/mol

Chemical Identifiers

CAS 54132-75-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013868
InChI Key DZSGDHNHQAJZCO-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isocyanate, 3,5-dimethylphenylisocyanate, 3,5-xylyl isocyanate, benzene, 1-isocyanato-3,5-dimethyl, isocyanic acid 3,5-dimethylphenyl ester, 5-isocyanato-m-xylene, 1-isocyanato-3,5-dimethyl-benzene, acmc-1axct, 3,5-dimethylbenzenisocyanate, ksc274i2t
PubChem CID 521488
IUPAC Name 1-isocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=O)C

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2206
Boiling Point 205°C
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 3,5-Dimethylphenyl Isocyanate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.