Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D339625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3396-25G 25 g
D33965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3396-25G Supplier TCI America Supplier No. D339625G
Add to Cart
Add to Cart

Specifications

CAS 172975-69-8
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C8H11BO2
MDL Number MFCD00185689
Synonym 3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl
InChI Key DJGHSJBYKIQHIK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)C)C)(O)O
IUPAC Name (3,5-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.984
PubChem CID 2734349
Formula Weight 149.98
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.