Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Dimethylphenylmagnesium Bromide Solution

Catalog No. 56207650ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-207-650ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 56-207-650ML Supplier Sigma Aldrich Supplier No. 56207650ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,5-Dimethylphenylmagnesium bromide solution can be used:As a starting material for the synthesis of diarylborinic acids, which on Suzuki coupling with vinyl triflate yield penultimate methyl ester.To prepare 3,5-dimethyl-p-terphenyl by reacting with biphenylsulfonate in the presence of dppfNiCl2 catalyst.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 56812-60-3
Density 0.770 g/mL at 25°C
Boiling Point 34.6°C
Molecular Formula C7H6Br2Mg
Linear Formula BrC6H4CH2MgBr
MDL Number MFCD01319903
Quantity 50 mL
Molecular Weight (g/mol) 274.24 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.