Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylpyrazole, 99%

Catalog No. p-4258978
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116701000 100 g Plastic bottle
AC116705000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116701000 Supplier Thermo Scientific Chemicals Supplier No. 116701000
Only null left
Add to Cart

CAS: 67-51-6 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-51-6
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005243
InChI Key SDXAWLJRERMRKF-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
PubChem CID 6210
IUPAC Name 3,5-dimethyl-1H-pyrazole
SMILES CC1=CC(C)=NN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 106.0°C to 109.0°C
Color White
Boiling Point 218.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 23,74
Solubility Information Solubility in water: soluble
Formula Weight 96.13
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3, 5-Dimethylpyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to
EINECSNumber 200-657-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.