Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitroaniline 98.0+%, TCI America™
SDP

Catalog No. D245825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D245825G 25 g
D24585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D245825G Supplier TCI America Supplier No. D245825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 618-87-1
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.123
MDL Number MFCD00007263
InChI Key MPBZUKLDHPOCLS-UHFFFAOYSA-N
PubChem CID 12068
IUPAC Name 3,5-dinitroaniline
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N

Specifications

Melting Point 162°C
Quantity 25 g
UN Number 1596
Synonym benzenamine, 3,5-dinitro, aniline, 3,5-dinitro, unii-0ahr6k1n73, ccris 3109, 3,5-dinitrophenylamine, 3-5-dinitro-phenylamine, 3,5-dinitroanilin, aniline,5-dinitro, acmc-209mx8, benzenamine,3,5-dinitro
Formula Weight 183.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitroaniline
RTECSNumber BX9200100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.