Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Dinitrobenzoyl Chloride

Catalog No. 156272100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-627-2100G 100 g
15-627-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-627-2100G Supplier Sigma Aldrich Supplier No. 156272100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,5-Dinitrobenzoyl Chloride

Specifications

CAS 99-33-2
Melting Point 68°C to 69°C
Boiling Point 196°C (11 mmHg, lit.)
Molecular Formula C7H3ClN2O5
Linear Formula (O2N)2C6H3COCl
MDL Number MFCD00007248
Quantity 100 g
Synonym DNBC; NSC 2697
Molecular Weight (g/mol) 230.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.