Learn More
Cayman Chemical 3,5-Dinitrocatechol, Purity: ≥98%, CAS: 7659-29-2, 10 mg
SDP
Catalog No. 502939669
Encompass_Preferred
A potent COMT inhibitor (IC50 = 12 nM); selective for COMT over tyrosine hydroxylase, dopamine β-hydroxylase, dopamine decarboxylase, MAO-A, and MAO-B (IC50s = >14 μM for all); reduces 3-OMD levels in rat serum when administered in combination with L-DOPA and carbidopa compared with control animals receiving only L-DOPA and carbidopa at 30 mg/kg; chronic administration induces COMT-dependent thermal hyperalgesia and increases preference for dark in a light/dark preference test in rats at 15 mg/kg per day for seven days
Catalog No. 50-293-9669 Supplier Cayman Chemical Supplier No. 2918010 MG
A potent COMT inhibitor (IC50 = 12 nM); selective for COMT over tyrosine hydroxylase, dopamine β-hydroxylase, dopamine decarboxylase, MAO-A, and MAO-B (IC50s = >14 μM for all); reduces 3-OMD levels in rat serum when administered in combination with L-DOPA and carbidopa compared with control animals receiving only L-DOPA and carbidopa at 30 mg/kg; chronic administration induces COMT-dependent thermal hyperalgesia and increases preference for dark in a light/dark preference test in rats at 15 mg/kg per day for seven days