Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrosalicylic acid, 97+%

Catalog No. aaa1698818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1698818 50 g
AAA1698830 250 g
AAA169880B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1698818 Supplier Thermo Scientific Chemicals Supplier No. A1698818
Only null left
Add to Cart

CAS: 609-99-4 | C7H4N2O7 | 228.116 g/mol

3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-99-4
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.116
MDL Number MFCD00007104
InChI Key LWFUFLREGJMOIZ-UHFFFAOYSA-N
Synonym 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid
PubChem CID 11873
ChEBI CHEBI:53648
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 169°C to 174°C
Quantity 50 g
Beilstein 2220661
Solubility Information Soluble in water,dimethyl sulfoxide,methanol,ethanol,diethyl ether and benzene.
Formula Weight 228.12
Percent Purity ≥97%
Chemical Name or Material 3,5-Dinitrosalicylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-204-3
RTECSNumber VO2965000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.