Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrosalicylic Acid, 98%, Spectrum™ Chemical
SDP

Catalog No. p-7157327 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
25 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-126 100 g Amber Glass Bottle
18-600-127 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-600-126 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2288100GM
Only null left
Add to Cart
Add to Cart

D2288, 609-99-4, C7H4N2O7

Spectrum™ Chemical 3,5-Dinitrosalicylic Acid , also known as DNS or DNSA, is used for quantifying carbohydrates levels in blood. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 609-99-4
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.12
InChI Key LWFUFLREGJMOIZ-UHFFFAOYSA-N
IUPAC Name 2-hydroxy-3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.