Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diphenyl-1-(4-biphenylyl)formazan 95.0+%, TCI America™
SDP

Catalog No. B0277100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0277100MG 100 mg
B02771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0277100MG Supplier TCI America Supplier No. B0277100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21520-85-4
Molecular Formula C25H20N4
Molecular Weight (g/mol) 376.463
MDL Number MFCD00059680
InChI Key LKFXJYZDDITMNN-UHFFFAOYSA-N
Synonym 3,5-Diphenyl-1-(p-diphenylyl)formazan
PubChem CID 53424949
IUPAC Name N'-anilino-N-(4-phenylphenyl)iminobenzenecarboximidamide
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N=NC(=NNC3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 169°C
Color Purple-Red
Quantity 100 mg
Formula Weight 376.46
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diphenyl-1-(4-biphenylyl)formazan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.