Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diphenyl-1-(p-tolyl)formazan 96.0+%, TCI America™
SDP

Catalog No. B02851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02851G 1 g
B0285100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02851G Supplier TCI America Supplier No. B02851G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 13412-07-2
Molecular Formula C20H18N4
Molecular Weight (g/mol) 314.39
MDL Number MFCD00059316
InChI Key TYLDKKLAWINVKP-UHFFFAOYSA-N
Synonym p-Tolyltetrazolium Red Formazan
PubChem CID 3494325
IUPAC Name N-[(4-methylphenyl)imino]-N'-(phenylamino)benzenecarboximidamide
SMILES CC1=CC=C(C=C1)N=NC(=NNC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 154°C
Quantity 1 g
Formula Weight 314.39
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diphenyl-1-(p-tolyl)formazan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.