Learn More
Medchemexpress LLC (3,5-diphenylphenyl) N-cyclopentylcarbamate | 1141802-49-4 | MFCD34567719 | 99.5% | C24H23NO2 | 10MG
SDP
Catalog No. 5000260498
Encompass_Preferred
LUF5771 is an allosteric modulator of the luteinizing hormone (LH) receptor that acts as a potent inhibitor of recombinant LH (recLH) and Org 43553 while exhibiting low-efficacy partial activation of the LH receptor. Supplied as a powdered research reagent (CAS 1141802-49-4; molecular formula C24H23NO2), it is intended for laboratory research use only.
  • Potent allosteric inhibitor of recLH and Org 43553.
  • Partial agonist activity at the LH receptor with low efficacy.
  • Provided as a powder suitable for research applications.
  • High reported purity (about 99.5%).
  • Characterized by CAS number 1141802-49-4 and formula C24H23NO2.
Catalog No. 50-002-60498 Supplier Medchemexpress LLC Supplier No. HY13930310MG
LUF5771 is an allosteric modulator of the luteinizing hormone (LH) receptor that acts as a potent inhibitor of recombinant LH (recLH) and Org 43553 while exhibiting low-efficacy partial activation of the LH receptor. Supplied as a powdered research reagent (CAS 1141802-49-4; molecular formula C24H23NO2), it is intended for laboratory research use only.
  • Potent allosteric inhibitor of recLH and Org 43553.
  • Partial agonist activity at the LH receptor with low efficacy.
  • Provided as a powder suitable for research applications.
  • High reported purity (about 99.5%).
  • Characterized by CAS number 1141802-49-4 and formula C24H23NO2.