Learn More
CHEMSCIENCE Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 3.5, Individually Serialized, Tolerance ± 0.01 mL
Catalog No. NC2318863
Encompass_Preferred
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 3.5, Individually Serialized, Tolerance ± 0.01 mL
Catalog No. NC2318863 Supplier CHEMSCIENCE Supplier No. 123.302.03A
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 3.5, Individually Serialized, Tolerance ± 0.01 mL