Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Pyrazoledicarboxylic Acid Monohydrate 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
303180-11-2
C5H6N2O5
174.11
MFCD00149323
GLINCONFUZIMCN-UHFFFAOYSA-N
3,5-pyrazoledicarboxylic acid,pyrazole-3,5-dicarboxylic acid,pyrazole-3,5-dicarboxylic acid monohydrate,3,5-pyrazol dicarboxylic acid,3,5-pyrazole dicarboxylic acid,zlchem 726,3,5-dicarboxypyrazole,acmc-20a0hc,ksc223c6j,3,5-pyrazole-dicarboxylic acid
76559
1H-pyrazole-3,5-dicarboxylic acid
O.OC(=O)C1=CC(=NN1)C(O)=O
303180-11-2
C5H6N2O5
174.11
MFCD00149323
GLINCONFUZIMCN-UHFFFAOYSA-N
3,5-pyrazoledicarboxylic acid,pyrazole-3,5-dicarboxylic acid,pyrazole-3,5-dicarboxylic acid monohydrate,3,5-pyrazol dicarboxylic acid,3,5-pyrazole dicarboxylic acid,zlchem 726,3,5-dicarboxypyrazole,acmc-20a0hc,ksc223c6j,3,5-pyrazole-dicarboxylic acid
76559
1H-pyrazole-3,5-dicarboxylic acid
O.OC(=O)C1=CC(=NN1)C(O)=O
Melting Point 292°C to 295°C
Water 11% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 25, 162
Solubility Information Solubility in water: insoluble
Formula Weight 174.11
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3, 5-Pyrazoledicarboxylic acid monohydrate, 97%
Show More Show Less

;Warning

EINECSNumber : 221-474-7

TSCA : TSCA