Learn More
Medchemexpress LLC 3,5-Pyridinedicarboxylic acid, 4-(2,3-dichlorophenyl)-1,4-dihydro-2,6-dimethyl-, 3-ethyl 5-methyl ester-d3 | 1219795-30-8 | 99.8% | 1 MG
SDP
Catalog No. 5000406898
Encompass_Preferred
Felodipine-d3 is a deuterium-labeled variant of Felodipine, a potent dihydropyridine calcium channel antagonist. It lowers blood pressure by selectively acting on vascular smooth muscle. This anti-hypertensive agent induces autophagy and can cross the blood-brain barrier. It is suitable for research use only and can be used as a tracer or an internal standard for quantitative analysis.
  • Potent vasoselective calcium channel antagonist
  • Lowers blood pressure by selective action on vascular smooth muscle
  • Induces autophagy
  • Can cross the blood-brain barrier
  • Suitable for research use only
  • Used as a tracer
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
Catalog No. 50-004-06898 Supplier Medchemexpress LLC Supplier No. HYB0309S21MG
Felodipine-d3 is a deuterium-labeled variant of Felodipine, a potent dihydropyridine calcium channel antagonist. It lowers blood pressure by selectively acting on vascular smooth muscle. This anti-hypertensive agent induces autophagy and can cross the blood-brain barrier. It is suitable for research use only and can be used as a tracer or an internal standard for quantitative analysis.
  • Potent vasoselective calcium channel antagonist
  • Lowers blood pressure by selective action on vascular smooth muscle
  • Induces autophagy
  • Can cross the blood-brain barrier
  • Suitable for research use only
  • Used as a tracer
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS