Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-Pyridinedicarboxylic acid, 4-(2,3-dichlorophenyl)-1,4-dihydro-2,6-dimethyl-, 3-methyl ester | 123853-39-4 | 95.1% | 356.20 | 1 G
SDP

Catalog No. 5000404023
Encompass_Preferred

Clevidipine butyrate impurity 1 is an impurity of Clevidipine butyrate, supplied as a solid. This product is for research use only and not for sale to patients.

  • High purity: 95.06%
  • Molecular weight: 356.20
  • Molecular formula: C16H15Cl2NO4
  • CAS number: 123853-39-4
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Classified as a drug intermediate

Catalog No. 50-004-04023 Supplier Medchemexpress LLC Supplier No. HY330131G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clevidipine butyrate impurity 1 is an impurity of Clevidipine butyrate, supplied as a solid. This product is for research use only and not for sale to patients.

  • High purity: 95.06%
  • Molecular weight: 356.20
  • Molecular formula: C16H15Cl2NO4
  • CAS number: 123853-39-4
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Classified as a drug intermediate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.