Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Pyridinedicarboxylic acid

Catalog No. P6420010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6420010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P6420010G Supplier Sigma Aldrich Supplier No. P6420010G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-phenyl-1-butanol is oxidized by ceric ammonium nitrate in acetonitrile to afford 2-phenyltetrahydrofuran
  • It undergoes cyclization in presence of phosphoric acid at high temperature to yield tetralin.4-phenyl-1-butanol was used in synthesis of nk105, a paclitaxel-incorporating micellar nanoparticle formulation

Specifications

CAS 499-81-0
Melting Point >300°C (lit.)
Molecular Formula C7H5NO4
Linear Formula C7H5NO4
MDL Number MFCD00006393
Quantity 10 g
Synonym Dinicotinic acid
Molecular Weight (g/mol) 167.12
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.