Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-pyridinedicarboxylic acid dimethyl ester derivative | 186185-03-5 | 99.9% | 25 MG
SDP

Catalog No. 5000234522
Encompass_Preferred

BMS-193885 is a selective neuropeptide Y1 (NPY Y1) receptor antagonist supplied as a 25 mg research-grade sample with high purity for use in biochemical and pharmacological studies.

  • High purity (99.9%).
  • Molecular formula C33H42N4O6.
  • Molecular weight 590.71 g/mol.
  • Provided as a 25 mg sample (solid).
  • Suitable for in vitro and in vivo studies of appetite regulation and NPY signaling.

Catalog No. 50-002-34522 Supplier Medchemexpress LLC Supplier No. HY12061925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-193885 is a selective neuropeptide Y1 (NPY Y1) receptor antagonist supplied as a 25 mg research-grade sample with high purity for use in biochemical and pharmacological studies.

  • High purity (99.9%).
  • Molecular formula C33H42N4O6.
  • Molecular weight 590.71 g/mol.
  • Provided as a 25 mg sample (solid).
  • Suitable for in vitro and in vivo studies of appetite regulation and NPY signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.