Learn More
Med Vet International 3.5 TPO/DPO Locking Plate 25 Left, J1211LA
Catalog No. 5000581411
Encompass_Preferred
Triple Pelvic Osteotomy (TPO) and Double Pelvic Osteotomy (DPO) remain effrective techniques for the managment of hip dysplasia. Use of diverging, locking screws increases plate security as does the sightly larger footprint of our plates. Available in both 2.7 and 3.5 locking screw sizes at the following angles: 20.25 and 30 degrees. Extra long locking drill guides are available. 3.5mm TPO/DPO Locking Plate 25 Left.
Catalog No. 50-005-81411 Supplier Med Vet International Supplier No. JOR1211LA
Add to Cart
Add to Cart
Triple Pelvic Osteotomy (TPO) and Double Pelvic Osteotomy (DPO) remain effrective techniques for the managment of hip dysplasia. Use of diverging, locking screws increases plate security as does the sightly larger footprint of our plates. Available in both 2.7 and 3.5 locking screw sizes at the following angles: 20.25 and 30 degrees. Extra long locking drill guides are available. 3.5mm TPO/DPO Locking Plate 25 Left.