Learn More
Cayman Chemical 3,5-Diiodothyroacetic Acid, Purity: ≥98%, CAS: 1155-40-4, 10 mg
SDP
Catalog No. 502987017
Encompass_Preferred
A thyroid hormone derivative; inhibits copper-induced lipid peroxidation of LDL isolated from human plasma at 1 µM; increases urinary and fecal excretion of sterols and prevents high-fat diet-induced increases in liver lipid and cholesterol levels, in rats at 5 mg/100 g diet
Catalog No. 50-298-7017 Supplier Cayman Chemical Supplier No. 2210810 MG
A thyroid hormone derivative; inhibits copper-induced lipid peroxidation of LDL isolated from human plasma at 1 µM; increases urinary and fecal excretion of sterols and prevents high-fat diet-induced increases in liver lipid and cholesterol levels, in rats at 5 mg/100 g diet