Learn More
Alkali Scientific 3.5mL Clear Glass 20x50mm High Recovery Vial with Graduations 20 400 Screw Thread. 3.3 Borosilicate, 12/Pk
SDP
Catalog No. 5000661930
Encompass_Preferred
The Alkali Scientific™ 3.5 mL High Recovery Vial is designed for accurate sample handling and maximum recovery in analytical and laboratory applications. Manufactured from high quality 3.3 borosilicate clear glass, this vial provides excellent chemical resistance and thermal stability while allowing clear visibility of the contents. The high recovery design helps minimize residual sample loss, making it suitable for applications where sample preservation is critical. Graduated markings allow convenient visual volume reference during preparation and transfer steps. Featuring a 20 400 screw thread finish, the vial is compatible with standard laboratory caps and closures for secure sealing. With dimensions of 20 × 50 mm, it fits common laboratory racks and autosampler setups used in analytical workflows. Supplied in packs of 12, these vials are well suited for routine analysis, sample storage, and precision laboratory applications.
Catalog No. 50-006-61930 Supplier Alkali Scientific Supplier No. HRV03.5
The Alkali Scientific™ 3.5 mL High Recovery Vial is designed for accurate sample handling and maximum recovery in analytical and laboratory applications. Manufactured from high quality 3.3 borosilicate clear glass, this vial provides excellent chemical resistance and thermal stability while allowing clear visibility of the contents. The high recovery design helps minimize residual sample loss, making it suitable for applications where sample preservation is critical. Graduated markings allow convenient visual volume reference during preparation and transfer steps. Featuring a 20 400 screw thread finish, the vial is compatible with standard laboratory caps and closures for secure sealing. With dimensions of 20 × 50 mm, it fits common laboratory racks and autosampler setups used in analytical workflows. Supplied in packs of 12, these vials are well suited for routine analysis, sample storage, and precision laboratory applications.