Learn More
Azzota Corp Disposable Uv-Vis Cuvettes - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm. Outer dimension of 12.5mm x 12.5mm x 45mm Methacrylate Pathlength: 10mm 3.5Ml Volume Wavelength Range: 285 To 800Nm (Uv-Vis) Package Of 100
Catalog No. 507532900
Encompass_Preferred
Disposable Uv-Vis Cuvettes - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm. Outer dimension of 12.5mm x 12.5mm x 45mm Methacrylate Pathlength: 10mm 3.5Ml Volume Wavelength Range: 285 To 800Nm (Uv-Vis) Package Of 100
Catalog No. 50-753-2900 Supplier Azzota Corp Supplier No. A907
Disposable Uv-Vis Cuvettes - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm. Outer dimension of 12.5mm x 12.5mm x 45mm Methacrylate Pathlength: 10mm 3.5Ml Volume Wavelength Range: 285 To 800Nm (Uv-Vis) Package Of 100