Learn More
Azzota Corp Disposable Vis Cuvettes - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm Polystyrene Pathlength: 10mm 3.5Ml Volume Wavelength Range: 340-800Nm (Vis) 100/Pk
Catalog No. 507532902
Encompass_Preferred
Disposable Vis Cuvettes - 3.5ml x 100 Square Cuvettes 10mm x 10mm x 45mm Polystyrene Pathlength: 10mm 3.5ml volume Wavelength Range: 340-800nm (Vis) Package of 100 Shipping Size: 9 x 9 x 3 inch Shipping Weight: 1 lbs
Catalog No. 50-753-2902 Supplier Azzota Corp Supplier No. A902
Disposable Vis Cuvettes - 3.5ml x 100 Square Cuvettes 10mm x 10mm x 45mm Polystyrene Pathlength: 10mm 3.5ml volume Wavelength Range: 340-800nm (Vis) Package of 100 Shipping Size: 9 x 9 x 3 inch Shipping Weight: 1 lbs