Learn More
Sarstedt Inc 3.5ML SCTUBE 13X92 FCBR/PK1000
Catalog No. NC9941656 Shop All Sarstedt Inc. Products
Encompass_Preferred
3.5ml Screen Printed Screw Cap Tube 13x92mm False Bottom Round Polypropylene Cap Assembled Graduations and Writing Block
Catalog No. NC9941656 Supplier Sarstedt Inc Supplier No. 62.617
3.5ml Screen Printed Screw Cap Tube 13x92mm False Bottom Round Polypropylene Cap Assembled Graduations and Writing Block