Learn More
Sarstedt Inc 3.5ml Screw Cap Tube, Conical Bottom, Polypropylene, 1000/Pack
Catalog No. NC9697779 Shop All Sarstedt Inc. Products
Encompass_Preferred
3.5ml Screw Cap Tube 11.5x66mm Conical Bottom Skirted Polypropylene Neutral Cap
Catalog No. NC9697779 Supplier Sarstedt Inc Supplier No. 60.549
3.5ml Screw Cap Tube 11.5x66mm Conical Bottom Skirted Polypropylene Neutral Cap