Learn More
Electron Microscopy Sciences 3.5x Auxiliary Lens, 1.75x
SDP
Catalog No. 5026458
Encompass_Preferred
Lightweight, adjustable headband for comfort provides close-up magnification while leaving both hands free for working. Tilt up and away when not in use. Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue. Auxillary lens to be used with 68108-05, 68108-06, or 68108-07.
Magnification: 1.75x
Catalog No. 50-264-58 Supplier Electron Microscopy Sciences Supplier No. 6810820
Lightweight, adjustable headband for comfort provides close-up magnification while leaving both hands free for working. Tilt up and away when not in use. Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue. Auxillary lens to be used with 68108-05, 68108-06, or 68108-07.
Magnification: 1.75x