Learn More
Med Vet International 3.5x Clip-On Surgical Loupes, J1138C
Supplier:  Med Vet International JOR1138C
Encompass_Preferred
Working Distance: 375±10mm ¥ Depth of Field: 20mm ¥ Field of View: 50mm ¥ Weight: 2.4 oz.
Catalog No. 50-005-83946