Learn More
Medchemexpress LLC 3,6,11-Trioxa-9-azatridecanoic acid, 12,12-dimethyl-10-oxo-. | 108466-89-3 | 263.29 | 10 G
SDP
Catalog No. 5000445476
Encompass_Preferred
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by this linker: one targeting an E3 ubiquitin ligase and the other for a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Essential for the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-004-45476 Supplier Medchemexpress LLC Supplier No. HYW00505610G
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by this linker: one targeting an E3 ubiquitin ligase and the other for a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Essential for the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system