Learn More
Medchemexpress LLC 3,6,11-Trioxa-9-azatridecanoic acid, 12,12-dimethyl-10-oxo- | 108466-89-3 | 263.29 | 25 G
SDP
Catalog No. 5000449859
Encompass_Preferred
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker
  • Includes a ligand for an E3 ubiquitin ligase
  • Includes a ligand for a target protein
Catalog No. 50-004-49859 Supplier Medchemexpress LLC Supplier No. 25GHYW005056
Add to Cart
Add to Cart
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker
  • Includes a ligand for an E3 ubiquitin ligase
  • Includes a ligand for a target protein