Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,11-Trioxa-9-azatridecanoic acid, 12,12-dimethyl-10-oxo- | 108466-89-3 | MFCD17019374 | 263.29 | 1 G
SDP

Catalog No. 5000445461
Encompass_Preferred

Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, to selectively degrade target proteins through the intracellular ubiquitin-proteasome system. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • For research use only
  • Molecular weight: 263.29
  • Formula: C11H21NO6
  • CAS no: 108466-89-3
  • Appearance: Colorless to light yellow liquid
  • Purity (LCMS): 99.87%

Catalog No. 50-004-45461 Supplier Medchemexpress LLC Supplier No. HYW0050561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, to selectively degrade target proteins through the intracellular ubiquitin-proteasome system. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • For research use only
  • Molecular weight: 263.29
  • Formula: C11H21NO6
  • CAS no: 108466-89-3
  • Appearance: Colorless to light yellow liquid
  • Purity (LCMS): 99.87%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.