Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontan-1-amine | 1977493-48-3 | >98.0% | 559.69 | C25H53NO12 | 50 MG
SDP

Catalog No. 5000202626
Encompass_Preferred

m-PEG12-amine is a methoxy-terminated polyethylene glycol (PEG) linker with a terminal primary amine used as a hydrophilic spacer for bioconjugation, PROTAC, and ADC linker synthesis. It provides aqueous solubility and a reactive amine for standard coupling chemistries.

  • Methoxy-terminated PEG12 spacer provides hydrophilicity.
  • Terminal primary amine enables standard coupling chemistry.
  • High purity: >98.0%.
  • Molecular weight approximately 559.69.
  • Appearance: colorless to off-white solid-liquid mixture.
  • Storage: store at -20°C, protect from light, under nitrogen.

Catalog No. 50-002-02626 Supplier Medchemexpress LLC Supplier No. HY14022750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG12-amine is a methoxy-terminated polyethylene glycol (PEG) linker with a terminal primary amine used as a hydrophilic spacer for bioconjugation, PROTAC, and ADC linker synthesis. It provides aqueous solubility and a reactive amine for standard coupling chemistries.

  • Methoxy-terminated PEG12 spacer provides hydrophilicity.
  • Terminal primary amine enables standard coupling chemistry.
  • High purity: >98.0%.
  • Molecular weight approximately 559.69.
  • Appearance: colorless to off-white solid-liquid mixture.
  • Storage: store at -20°C, protect from light, under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.