Learn More
Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30,33-Undecaoxapentatriacontan-1-amine, 35-azido- | 1800414-71-4 | MFCD08274605 | >98.0% | C24H50N4O11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140218100MG
Encompass_Preferred
Azido-PEG11-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group and an amine group. The product appears as a colorless to light yellow liquid and is intended for research use only, not for medical applications.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group and an amine group
  • Colorless to light yellow liquid
Catalog No. 50-003-57285