Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30-decaoxadotriacontane-1,32-diamine | 928292-69-7 | ≥97.0% | 500.62 | C22H48N2O10 | 100 MG
SDP

Catalog No. 5000209333
Encompass_Preferred

Amino-PEG10-amine is a polyethylene glycol (PEG) based diamine reagent used as a hydrophilic spacer and linker in PROTAC synthesis and other bioconjugation applications. It provides terminal primary amines for coupling to ligands or functional groups.

  • Primary diamine with two terminal amino groups.
  • Peg10 spacer provides hydrophilicity and flexibility.
  • Molecular weight approximately 500.62 g/mol.
  • CAS number 928292-69-7 for unambiguous identification.
  • Purity ≥97.0% by NMR, per certificate of analysis.
  • Suitable for linker construction and bioconjugation in synthetic chemistry workflows.

Catalog No. 50-002-09333 Supplier Medchemexpress LLC Supplier No. HYW040270100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG10-amine is a polyethylene glycol (PEG) based diamine reagent used as a hydrophilic spacer and linker in PROTAC synthesis and other bioconjugation applications. It provides terminal primary amines for coupling to ligands or functional groups.

  • Primary diamine with two terminal amino groups.
  • Peg10 spacer provides hydrophilicity and flexibility.
  • Molecular weight approximately 500.62 g/mol.
  • CAS number 928292-69-7 for unambiguous identification.
  • Purity ≥97.0% by NMR, per certificate of analysis.
  • Suitable for linker construction and bioconjugation in synthetic chemistry workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.