Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30-decaoxadotriacontane-1,32-diamine | 928292-69-7 | MFCD28122962 | ≥97.0% | 500.62 g/mol | C22H48N2O10 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYW040270

Encompass_Preferred

Amino-PEG10-amine is a polyethylene glycol (PEG10) diamine linker used in PROTAC synthesis to connect ligands and improve solubility and flexibility. It is supplied as a high-purity chemical building block for medicinal chemistry and linker design.

  • PEG10 diamine linker for PROTAC synthesis.
  • Molecular weight 500.62 g/mol.
  • Chemical formula C22H48N2O10.
  • Purity ≥97.0% (NMR) as per COA.
  • Appearance: colorless to light yellow, solid-liquid mixture.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09603


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.