Learn More
Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30-decaoxadotriacontane-1,32-diamine | 928292-69-7 | MFCD28122962 | ≥97.0% | 500.62 g/mol | C22H48N2O10 | 50 MG
SDP
Catalog No. 5000208931
Encompass_Preferred
Amino-PEG10-amine is a polyethylene glycol (PEG10) diamine linker bearing primary amino groups at both termini. It is used as a hydrophilic, flexible spacer for PROTAC synthesis, bioconjugation, and crosslinking, providing improved solubility and coupling handle accessibility.
  • Provides two terminal primary amines for covalent coupling.
  • PEG10 spacer increases solubility in aqueous media.
  • High purity suitable for research and synthesis (≥97% by NMR).
  • Flexible spacer for linker design in PROTAC and bioconjugation applications.
  • Stable when stored protected from light at recommended temperatures.
Catalog No. 50-002-08931 Supplier Medchemexpress LLC Supplier No. HYW04027050MG
Amino-PEG10-amine is a polyethylene glycol (PEG10) diamine linker bearing primary amino groups at both termini. It is used as a hydrophilic, flexible spacer for PROTAC synthesis, bioconjugation, and crosslinking, providing improved solubility and coupling handle accessibility.
  • Provides two terminal primary amines for covalent coupling.
  • PEG10 spacer increases solubility in aqueous media.
  • High purity suitable for research and synthesis (≥97% by NMR).
  • Flexible spacer for linker design in PROTAC and bioconjugation applications.
  • Stable when stored protected from light at recommended temperatures.