Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21,24-octaoxapentacosan-1-amine | 869718-81-0 | MFCD13184961 | 98.0% | 383.48 g/mol | C17H37NO8 | 10 G
SDP

Catalog No. 5000203211
Encompass_Preferred

m-PEG8-Amine is a monomethyl polyethylene glycol (PEG8) primary amine used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and other bioconjugation applications. It provides an eight-unit PEG spacer with a terminal primary amine for conjugation chemistry, and is supplied at high purity for research use.

  • Cleavable PEG8 linker suitable for ADC and bioconjugation chemistry.
  • Provides an eight-unit (PEG8) flexible spacer with a terminal primary amine.
  • High purity suitable for research use: 98.0%.
  • Molecular weight approximately 383.48 g/mol; formula C17H37NO8.
  • SMILES COCCOCCOCCOCCOCCOCCOCCOCCN; compatible with common conjugation reactions.

Catalog No. 50-002-03211 Supplier Medchemexpress LLC Supplier No. HYW04023610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG8-Amine is a monomethyl polyethylene glycol (PEG8) primary amine used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and other bioconjugation applications. It provides an eight-unit PEG spacer with a terminal primary amine for conjugation chemistry, and is supplied at high purity for research use.

  • Cleavable PEG8 linker suitable for ADC and bioconjugation chemistry.
  • Provides an eight-unit (PEG8) flexible spacer with a terminal primary amine.
  • High purity suitable for research use: 98.0%.
  • Molecular weight approximately 383.48 g/mol; formula C17H37NO8.
  • SMILES COCCOCCOCCOCCOCCOCCOCCOCCN; compatible with common conjugation reactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.