Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21,24-octaoxapentacosan-1-amine | 869718-81-0 | MFCD13184961 | ≥98.0% | 383.48 g/mol | C17H37NO8 | 5 G
SDP

Catalog No. 5000203216
Encompass_Preferred

m-PEG8-Amine is a polyethylene glycol (PEG) based cleavable linker terminating in a primary amine, used for bioconjugation and the synthesis of antibody-drug conjugates. It provides an eight-unit PEG spacer to improve solubility and flexibility when coupling payloads to biomolecules.

  • Primary amine-terminated PEG spacer (8 units)
  • Cleavable linker suitable for bioconjugation and ADC synthesis
  • High purity: ≥98.0% (NMR)
  • Molecular weight: 383.48 g/mol
  • Chemical formula: C17H37NO8
  • CAS number: 869718-81-0
  • Storage: store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-03216 Supplier Medchemexpress LLC Supplier No. HYW0402365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG8-Amine is a polyethylene glycol (PEG) based cleavable linker terminating in a primary amine, used for bioconjugation and the synthesis of antibody-drug conjugates. It provides an eight-unit PEG spacer to improve solubility and flexibility when coupling payloads to biomolecules.

  • Primary amine-terminated PEG spacer (8 units)
  • Cleavable linker suitable for bioconjugation and ADC synthesis
  • High purity: ≥98.0% (NMR)
  • Molecular weight: 383.48 g/mol
  • Chemical formula: C17H37NO8
  • CAS number: 869718-81-0
  • Storage: store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.