Learn More
Medchemexpress LLC 3,6,9,12,15,18,21-Heptaoxatetracos-23-yn-1-ol | 1422023-54-8 | ≥97.0% | 1 G
SDP
Catalog No. 5000339857
Encompass_Preferred
Propargyl-PEG7-alcohol is a versatile PEG linker with the chemical name 3,6,9,12,15,18,21-Heptaoxatetracos-23-yn-1-ol. It has a molecular formula of C17H32O8 and a molecular weight of 364.43. This product is commonly utilized in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • High purity of ≥97.0%
  • Appears as a colorless to light yellow liquid
  • Suitable for ADC and PROTAC development
  • Stable storage conditions provided
Catalog No. 50-003-39857 Supplier Medchemexpress LLC Supplier No. HY1303781G
Propargyl-PEG7-alcohol is a versatile PEG linker with the chemical name 3,6,9,12,15,18,21-Heptaoxatetracos-23-yn-1-ol. It has a molecular formula of C17H32O8 and a molecular weight of 364.43. This product is commonly utilized in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • High purity of ≥97.0%
  • Appears as a colorless to light yellow liquid
  • Suitable for ADC and PROTAC development
  • Stable storage conditions provided