Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15,18,21-Heptaoxatetracos-23-yn-1-ol | 1422023-54-8 | ≥97.0% | 250 MG
SDP

Catalog No. 5000341537
Encompass_Preferred

Propargyl-PEG7-alcohol is a chemical compound identified by CAS number 1422023-54-8. It has a molecular formula of C17H32O8 and a molecular weight of 364.43. This compound is typically found as a colorless to light yellow liquid, with a purity of ≥97.0%. It is intended for research use only.

  • Molecular formula: C17H32O8
  • Molecular weight: 364.43
  • Purity: ≥97.0%
  • Appearance: colorless to light yellow liquid
  • Storage for pure form: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Catalog No. 50-003-41537 Supplier Medchemexpress LLC Supplier No. 250MGHY130378
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG7-alcohol is a chemical compound identified by CAS number 1422023-54-8. It has a molecular formula of C17H32O8 and a molecular weight of 364.43. This compound is typically found as a colorless to light yellow liquid, with a purity of ≥97.0%. It is intended for research use only.

  • Molecular formula: C17H32O8
  • Molecular weight: 364.43
  • Purity: ≥97.0%
  • Appearance: colorless to light yellow liquid
  • Storage for pure form: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.