Learn More
Medchemexpress LLC 3,6,9,12,15,18-Hexaoxanonadecan-1-amine | 184357-46-8 | ≥97.0% | 10 G
SDP
Catalog No. 5000340834
Encompass_Preferred
m-PEG6-Amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, while ADCs are comprised of an antibody to which is attached an ADC cytotoxin through an ADC linker.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Used in the synthesis of PROTACs
  • Used in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-003-40834 Supplier Medchemexpress LLC Supplier No. HY13040810G
m-PEG6-Amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, while ADCs are comprised of an antibody to which is attached an ADC cytotoxin through an ADC linker.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Used in the synthesis of PROTACs
  • Used in the synthesis of antibody-drug conjugates (ADCs)