Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecan-1-amine | 184357-46-8 | 97.0% | 295.37 g/mol | C13H29NO6 | 100 MG
SDP
Catalog No. 5000203916
Encompass_Preferred
m-PEG6-Amine is a methoxy-terminated PEG6 linker bearing a primary amine; it is used as a spacer and conjugation handle in PROTAC and antibody-drug conjugate (ADC) synthesis. The compound has formula C13H29NO6, molecular weight 295.37 g/mol, and is supplied at ~97.0% purity for research use.
  • PEG6 spacer with methoxy terminus and primary amine for conjugation
  • Suitable for PROTAC linker and ADC linker synthesis
  • Hydrophilic polyethylene glycol chain improves solubility and flexibility
  • Available in multiple pack sizes, including 100 MG
  • Provided at research-grade purity for synthetic applications
Catalog No. 50-002-03916 Supplier Medchemexpress LLC Supplier No. HY130408100MG
m-PEG6-Amine is a methoxy-terminated PEG6 linker bearing a primary amine; it is used as a spacer and conjugation handle in PROTAC and antibody-drug conjugate (ADC) synthesis. The compound has formula C13H29NO6, molecular weight 295.37 g/mol, and is supplied at ~97.0% purity for research use.
  • PEG6 spacer with methoxy terminus and primary amine for conjugation
  • Suitable for PROTAC linker and ADC linker synthesis
  • Hydrophilic polyethylene glycol chain improves solubility and flexibility
  • Available in multiple pack sizes, including 100 MG
  • Provided at research-grade purity for synthetic applications