Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecan-1-ol | 23601-40-3 | MFCD00045996 | 98.8% | 296.36 | C13H28O7 | 100 G
SDP
Catalog No. 5000210767
Encompass_Preferred
Hexaethylene glycol monomethyl ether is a polyethylene glycol (PEG) monomethyl ether used as a short, flexible linker and solubilizing spacer in synthetic and medicinal chemistry, notably in PROTAC construction.
  • Peg-based monomethyl ether providing a six-unit ethylene glycol spacer.
  • Improves solubility and flexibility of conjugates.
  • Suitable for incorporation into PROTACs and other bioconjugates.
  • High purity appropriate for synthetic chemistry applications.
  • Available in multiple package sizes, including 100 g.
Catalog No. 50-002-10767 Supplier Medchemexpress LLC Supplier No. HYW042625100G
Hexaethylene glycol monomethyl ether is a polyethylene glycol (PEG) monomethyl ether used as a short, flexible linker and solubilizing spacer in synthetic and medicinal chemistry, notably in PROTAC construction.
  • Peg-based monomethyl ether providing a six-unit ethylene glycol spacer.
  • Improves solubility and flexibility of conjugates.
  • Suitable for incorporation into PROTACs and other bioconjugates.
  • High purity appropriate for synthetic chemistry applications.
  • Available in multiple package sizes, including 100 g.