Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecan-1-ol | 23601-40-3 | MFCD00045996 | 98.8% | 296.36 g/mol | C13H28O7 | 10 G
SDP
Catalog No. 5000210654
Encompass_Preferred
Hexaethylene glycol monomethyl ether is a PEG-based monomethyl ether used as a linker and spacer in medicinal chemistry and PROTAC synthesis. It is supplied as a clear liquid with a molecular weight of 296.36 g/mol and high analytical purity for research applications.
  • High purity (98.8%)
  • Molecular formula C13H28O7
  • Suitable as a PEG linker for PROTAC and medicinal chemistry synthesis
  • Available in 10 G packaging
  • Liquid form, miscible with common organic solvents
Catalog No. 50-002-10654 Supplier Medchemexpress LLC Supplier No. HYW04262510G
Hexaethylene glycol monomethyl ether is a PEG-based monomethyl ether used as a linker and spacer in medicinal chemistry and PROTAC synthesis. It is supplied as a clear liquid with a molecular weight of 296.36 g/mol and high analytical purity for research applications.
  • High purity (98.8%)
  • Molecular formula C13H28O7
  • Suitable as a PEG linker for PROTAC and medicinal chemistry synthesis
  • Available in 10 G packaging
  • Liquid form, miscible with common organic solvents