Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecan-1-ol | 23601-40-3 | MFCD00045996 | 98.8% | 296.36 g·mol⁻¹ | C13H28O7 | 25 G
SDP
Catalog No. 5000208911
Encompass_Preferred
Hexaethylene glycol monomethyl ether is a methoxy-terminated polyethylene glycol (PEG) linker used as a hydrophilic spacer in the synthesis of PROTACs and other bifunctional molecules. It is a colorless to light yellow liquid with a molecular weight of 296.36 g·mol⁻¹ and high GC purity.
  • Methoxy-terminated hexaethylene glycol spacer
  • Hydrophilic and flexible linker suitable for PROTAC synthesis
  • High GC purity for research applications
  • Molecular weight of 296.36 g·mol⁻¹
  • Available in multiple pack sizes including 25 g
  • Colorless to light yellow liquid appearance
Catalog No. 50-002-08911 Supplier Medchemexpress LLC Supplier No. 25GHYW042625
Hexaethylene glycol monomethyl ether is a methoxy-terminated polyethylene glycol (PEG) linker used as a hydrophilic spacer in the synthesis of PROTACs and other bifunctional molecules. It is a colorless to light yellow liquid with a molecular weight of 296.36 g·mol⁻¹ and high GC purity.
  • Methoxy-terminated hexaethylene glycol spacer
  • Hydrophilic and flexible linker suitable for PROTAC synthesis
  • High GC purity for research applications
  • Molecular weight of 296.36 g·mol⁻¹
  • Available in multiple pack sizes including 25 g
  • Colorless to light yellow liquid appearance