Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecanoic acid | 16142-03-3 | 99.9% | 310.34 | C13H26O8 | 250 MG
SDP
Catalog No. 5000202195
Encompass_Preferred
m-PEG5-CH2COOH is a methyl-terminated PEG5 linker bearing a terminal carboxylic acid, designed for use in antibody-drug conjugate (ADC) and PROTAC linker synthesis as well as general bioconjugation and medicinal chemistry research.
  • High purity (99.9%).
  • Methyl-terminated PEG5 chain with a terminal carboxylic acid functional group.
  • Suitable for ADC and PROTAC linker synthesis and general bioconjugation.
  • Soluble in DMSO at 200 mg/mL; sonication recommended.
  • Stable when stored sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Supplied in research-scale packaging suitable for synthetic workflows.
Catalog No. 50-002-02195 Supplier Medchemexpress LLC Supplier No. HY120537250MG
m-PEG5-CH2COOH is a methyl-terminated PEG5 linker bearing a terminal carboxylic acid, designed for use in antibody-drug conjugate (ADC) and PROTAC linker synthesis as well as general bioconjugation and medicinal chemistry research.
  • High purity (99.9%).
  • Methyl-terminated PEG5 chain with a terminal carboxylic acid functional group.
  • Suitable for ADC and PROTAC linker synthesis and general bioconjugation.
  • Soluble in DMSO at 200 mg/mL; sonication recommended.
  • Stable when stored sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Supplied in research-scale packaging suitable for synthetic workflows.