Learn More
Medchemexpress LLC 3,6,9,12,15-pentaoxaheptadecan-1-ol, 17-azido-, 1-methanesulfonate | 352439-38-4 | 95.3% | C13H27N3O8S | 10MG
SDP
Catalog No. 5000261609
Encompass_Preferred
Azido-PEG6-MS is an azide-terminated polyethylene glycol (PEG6) mesylate linker supplied as a solid for use in PROTAC construction and general bioconjugation. It serves as a hydrophilic spacer with a terminal azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition, enabling modular assembly of bifunctional molecules and ease of downstream conjugation.
  • Provides an azide functional group for CuAAC and SPAAC reactions.
  • PEG6 spacer increases aqueous solubility and reduces aggregation.
  • Mesylate leaving group enables straightforward synthetic modification.
  • Available in small mg-scale quantities suitable for discovery research.
  • High purity appropriate for chemical synthesis applications.
Catalog No. 50-002-61609 Supplier Medchemexpress LLC Supplier No. HY13836310MG
Azido-PEG6-MS is an azide-terminated polyethylene glycol (PEG6) mesylate linker supplied as a solid for use in PROTAC construction and general bioconjugation. It serves as a hydrophilic spacer with a terminal azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition, enabling modular assembly of bifunctional molecules and ease of downstream conjugation.
  • Provides an azide functional group for CuAAC and SPAAC reactions.
  • PEG6 spacer increases aqueous solubility and reduces aggregation.
  • Mesylate leaving group enables straightforward synthetic modification.
  • Available in small mg-scale quantities suitable for discovery research.
  • High purity appropriate for chemical synthesis applications.