Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15-Pentaoxaheptadecane-1,17-diamine | 72236-26-1 | 97.00% | 25 G
SDP

Catalog No. 5000341977
Encompass_Preferred

Amino-PEG5-amine is a PEG-based (5 units) PROTAC linker. It is designed for use in the synthesis of PROTACs and is intended for research purposes only.

  • PEG-based (5 units) PROTAC linker
  • Utilized in the synthesis of PROTACs
  • For research use only
  • Appearance: colorless to light yellow liquid
  • Soluble in water: ≥ 100 mg/mL
  • Molecular weight: 280.36
  • Molecular formula: C12H28N2O5
  • Store at 4°C, protect from light
  • In solvent: store at -80°C for 6 months, or -20°C for 1 month, protect from light

Catalog No. 50-003-41977 Supplier Medchemexpress LLC Supplier No. 25GHY130447
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG5-amine is a PEG-based (5 units) PROTAC linker. It is designed for use in the synthesis of PROTACs and is intended for research purposes only.

  • PEG-based (5 units) PROTAC linker
  • Utilized in the synthesis of PROTACs
  • For research use only
  • Appearance: colorless to light yellow liquid
  • Soluble in water: ≥ 100 mg/mL
  • Molecular weight: 280.36
  • Molecular formula: C12H28N2O5
  • Store at 4°C, protect from light
  • In solvent: store at -80°C for 6 months, or -20°C for 1 month, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.