Learn More
Medchemexpress LLC 3,6,9,12,15-Pentaoxaoctadec-17-ynoic acid | 1429934-37-1 | ≥97.0% | 290.31 | 25 MG
SDP
Catalog No. 5000351391
Encompass_Preferred
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker and a click chemistry reagent. It contains an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Azide group-containing molecules. This linker is crucial for synthesizing PROTACs, which utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Enables targeted protein degradation
Catalog No. 50-003-51391 Supplier Medchemexpress LLC Supplier No. 25MGHY140025
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker and a click chemistry reagent. It contains an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Azide group-containing molecules. This linker is crucial for synthesizing PROTACs, which utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Enables targeted protein degradation