Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecan-1-ol, 14-bromo- | 957205-14-0 | 98.0% | C10H21BrO5 | 100 MG
SDP
Catalog No. 5000353189
Encompass_Preferred
Bromo-PEG5-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-53189 Supplier Medchemexpress LLC Supplier No. HY141214100MG
Bromo-PEG5-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system